KEY466133911 Key Organics / BIONET

(E)-1-[(1-chloro-3,4-dihydronaphthalen-2-yl)methylidene]-2-(4-methylphenyl)hydrazine

  • MDL number MFCD01569197
  • Cas Number : 1351286-49-1
  • Molecular Weight : 296.80

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9195

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12N-505S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart