KEY466133914 Key Organics / BIONET

{4-[(4-methylphenyl)sulfanyl]-3-nitrophenyl}methyl thiophene-2-carboxylate

  • MDL number MFCD01569202
  • Cas Number : 320424-07-5
  • Molecular Weight : 385.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9210

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12N-643S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart