KEY466133921 Key Organics / BIONET

3-[(2-chloro-6-fluorophenyl)methyl]-4-hydroxy-1-[2-(pyridin-2-yl)ethyl]-1,2-dihydropyridin-2-one

  • MDL number MFCD01871444
  • Cas Number : 477860-56-3
  • Molecular Weight : 358.80

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9244

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12N-721_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart