KEY466133923 Key Organics / BIONET

ethyl 3-(2-chloro-6-fluorophenyl)-2-[1-(6-fluoropyridin-2-yl)-1H-pyrazol-5-yl]propanoate

  • MDL number MFCD01871447
  • Cas Number : 477709-64-1
  • Molecular Weight : 391.80

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9253

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12N-733_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart