KEY466133925 Key Organics / BIONET

3-(3,5-dibromo-1H-1,2,4-triazol-1-yl)butan-2-one

  • MDL number MFCD01568885
  • Cas Number : 320424-38-2
  • Molecular Weight : 296.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9255

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12N-736_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart