KEY466133926 Key Organics / BIONET

7-chloro-N-[(4-fluorophenyl)methyl]quinazolin-4-amine

  • MDL number MFCD01815388
  • Cas Number : 477861-93-1
  • Molecular Weight : 287.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9280

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12N-822_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart