KEY466133931 Key Organics / BIONET

2-hydroxy-5-methylbenzene-1,3-dicarbonitrile

  • MDL number MFCD01568890
  • Cas Number : 73289-57-3
  • Molecular Weight : 158.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9302

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12N-951_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart