KEY466133932 Key Organics / BIONET

2-(4-tert-butylphenyl)-5-{[(4-nitrophenyl)methyl]sulfanyl}-1,3,4-oxadiazole

  • MDL number MFCD02082788
  • Cas Number : 477862-10-5
  • Molecular Weight : 369.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9308

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12P-003_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart