KEY466133933 Key Organics / BIONET

5-({4-[(2-chloro-6-fluorophenyl)methoxy]phenyl}methylidene)-1,3-diazinane-2,4,6-trione

  • MDL number MFCD02082790
  • Cas Number : 430469-95-7
  • Molecular Weight : 374.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9315

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12P-011_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart