KEY466133937 Key Organics / BIONET

(Z)-(ethyl N-[(Z)-benzoyl]ethanecarbohydrazonate)

  • MDL number MFCD02082701
  • Cas Number : 85936-50-1
  • Molecular Weight : 206.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9324

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12P-022_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart