KEY466133942 Key Organics / BIONET

2-cyano-N'-[(1E)-4,4,4-trifluoro-1-(furan-2-yl)-3-oxobutylidene]acetohydrazide

  • MDL number MFCD02082797
  • Cas Number : 477862-30-9
  • Molecular Weight : 287.198

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9339

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12P-043_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart