KEY466133945 Key Organics / BIONET

4-[4-(thiophen-2-yl)pyrimidin-2-yl]phenyl 3-methylbenzoate

  • MDL number MFCD02082800
  • Cas Number : 477862-35-4
  • Molecular Weight : 372.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9349

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12P-058_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart