KEY466133963 Key Organics / BIONET

1-ethyl-1H,2H,3H,4H,9H-pyrido[3,4-b]indole

  • MDL number MFCD01566864
  • Cas Number : 6678-86-0
  • Molecular Weight : 200.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9410

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12P-144_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart