KEY466133967 Key Organics / BIONET

4-oxo-4H,5H,6H,7H,8H-pyrazolo[1,5-a][1,4]diazepine-2-carboxylic acid

  • MDL number MFCD02570781
  • Cas Number : 477863-08-4
  • Molecular Weight : 195.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9418

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12P-158_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart