KEY466133969 Key Organics / BIONET

5-(thiophen-2-yl)-4-[(E)-[(thiophen-2-yl)methylidene]amino]-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD02102329
  • Cas Number : 477863-25-5
  • Molecular Weight : 292.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9427

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12P-307S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart