KEY466133978 Key Organics / BIONET

5-methoxy-2-phenyl-N-(prop-2-en-1-yl)pyrimidin-4-amine

  • MDL number MFCD02102339
  • Cas Number : 477863-87-9
  • Molecular Weight : 241.294

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9481

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12P-518S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart