KEY466133979 Key Organics / BIONET

methyl 3-[2-(methylcarbamoyl)-1H-pyrrol-1-yl]thiophene-2-carboxylate

  • MDL number MFCD02102340
  • Cas Number : 477863-90-4
  • Molecular Weight : 264.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9484

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12P-521S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart