KEY466133981 Key Organics / BIONET

tert-butyl 4-[(2,4-dichlorophenyl)amino]piperidine-1-carboxylate

  • MDL number MFCD04124518
  • Cas Number : 860785-94-0
  • Molecular Weight : 345.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9509

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12P-572S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart