KEY466133984 Key Organics / BIONET

2'-(2-fluorophenyl)-2,4-dihydrospiro[1-benzopyran-3,1'-cyclopropane]-4-one

  • MDL number MFCD02082710
  • Cas Number : 477864-18-9
  • Molecular Weight : 268.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9522

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12P-603_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart