KEY466133994 Key Organics / BIONET

bis(pyrazin-2-yl)-4H-1,2,4-triazol-4-amine

  • MDL number MFCD02570786
  • Cas Number : 223645-71-4
  • Molecular Weight : 240.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9547

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12P-644_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart