KEY466133995 Key Organics / BIONET

(4E)-4-[(thiophen-2-yl)methylidene]-2,3,4,5-tetrahydro-1-benzothiepin-5-one

  • MDL number MFCD02082836
  • Cas Number : 924275-80-9
  • Molecular Weight : 272.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9548

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12P-648_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart