KEY466134008 Key Organics / BIONET

N-[(cyclohexylcarbamoyl)amino]-1-(6-methylpyridin-2-yl)-1H-imidazole-4-carboxamide

  • MDL number MFCD02570859
  • Cas Number : 477864-45-2
  • Molecular Weight : 342.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9573

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12P-819_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart