KEY466134011 Key Organics / BIONET

2-[3-(2,5-dimethyl-1H-pyrrol-1-yl)thiophen-2-yl]-5-methylpyrazolo[1,5-a]pyrimidin-7-amine

  • MDL number MFCD03102514
  • Cas Number : 691869-93-9
  • Molecular Weight : 323.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9580

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12R-0011_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart