KEY466134014 Key Organics / BIONET

2-[(2H-1,3-benzodioxol-5-yl)methylidene]-2,3-dihydro-1H-indene-1,3-dione

  • MDL number MFCD00129358
  • Cas Number : 29874-34-8
  • Molecular Weight : 278.263

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9595

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12R-0206_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart