KEY466134015 Key Organics / BIONET

1-[3-(4-methylphenyl)-2,1-benzoxazol-5-yl]ethan-1-one

  • MDL number MFCD02661643
  • Cas Number : 439094-63-0
  • Molecular Weight : 251.285

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9599

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12R-0214_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart