KEY466134022 Key Organics / BIONET

4-(2H-1,3-benzodioxol-5-yl)-2-methyl-5-oxo-1H,4H,5H-indeno[1,2-b]pyridine-3-carbonitrile

  • MDL number MFCD03102685
  • Cas Number : 691870-02-7
  • Molecular Weight : 342.354

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9621

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12R-0261_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart