KEY466134027 Key Organics / BIONET

2,2-dimethyl-6-nitro-3,4-dihydro-2H-1,4-benzothiazine

  • MDL number MFCD06496126
  • Cas Number : 591221-28-2
  • Molecular Weight : 224.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63080

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12X-0955_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart