KEY466134030 Key Organics / BIONET

ethyl 4-(2-bromoacetyl)-5-methyl-1,2-oxazole-3-carboxylate

  • MDL number MFCD02661839
  • Cas Number : 115697-68-2
  • Molecular Weight : 276.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Oil
Purity 0.90
Storage temp. Store under N2
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64288

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404,P422
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1K-005_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart