KEY466134036 Key Organics / BIONET

4-[2-(4-chlorophenyl)-4-methyl-1,3-thiazol-5-yl]-N-(4-methyl-1,3-thiazol-2-yl)pyrimidin-2-amine

  • MDL number MFCD28347906
  • Cas Number : 1881329-72-1
  • Molecular Weight : 399.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 301 - 303
Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/453052

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2R-0838_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart