KEY466134037 Key Organics / BIONET

methyl 4-benzyl-6-(4-chlorophenyl)-1,3-dioxo-hexahydro-1H-furo[3,4-c]pyrrole-4-carboxylate

  • MDL number MFCD04125018
  • Cas Number : 1005085-90-4
  • Molecular Weight : 399.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69924

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2X-0844_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart