KEY466134039 Key Organics / BIONET

N-[(4-methylphenyl)methyl]methanesulfonamide

  • MDL number MFCD01213927
  • Cas Number : 42060-28-6
  • Molecular Weight : 199.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72061

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3N-017_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart