KEY466134043 Key Organics / BIONET

7-fluoro-1-[(4-methylphenyl)methyl]-4-oxo-1,4-dihydroquinoline-3-carbohydrazide

  • MDL number MFCD02083115
  • Cas Number : 478043-11-7
  • Molecular Weight : 325.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75648

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4M-333S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart