KEY466134065 Key Organics / BIONET

2-(benzyloxy)-6-fluorobenzonitrile

  • MDL number MFCD00068211
  • Cas Number : 94088-45-6
  • Molecular Weight : 227.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(benzyloxy)-6-fluorobenzonitrile
Inchi Code 1S/C14H10FNO/c15-13-7-4-8-14(12(13)9-16)17-10-11-5-2-1-3-6-11/h1-8H,10H2
Inchi Key ITKHCBLEEGSDOF-UHFFFAOYSA-N

Description

Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/358430

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P411,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/BH-0417_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart