KEY466134125 Key Organics / BIONET

N-[4-cyano-3-(trifluoromethyl)phenyl]acetamide

  • MDL number MFCD00052360
  • Cas Number : 97760-99-1
  • Molecular Weight : 228.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp Solid
Iupac Name N-[4-cyano-3-(trifluoromethyl)phenyl]acetamide
Inchi Code 1S/C10H7F3N2O/c1-6(16)15-8-3-2-7(5-14)9(4-8)10(11,12)13/h2-4H,1H3,(H,15,16)
Inchi Key FHKLGEQKLMMVCZ-UHFFFAOYSA-N

Description

Melting Point Solid
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/402985

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/FS-1072_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart