KEY466134147 Key Organics / BIONET

1-(5-bromo-2-methoxyphenyl)ethan-1-one

  • MDL number MFCD03789141
  • Cas Number : 16740-73-1
  • Molecular Weight : 229.073

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/403487

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/FS-1584_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart