KEY466134155 Key Organics / BIONET

(2-fluoro-6-methoxyphenyl)methanamine

  • MDL number MFCD00042292
  • Cas Number : 150517-75-2
  • Molecular Weight : 155.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-fluoro-6-methoxyphenyl)methanamine
Inchi Code 1S/C8H10FNO/c1-11-8-4-2-3-7(9)6(8)5-10/h2-4H,5,10H2,1H3
Inchi Key JCXMQSDLDAJYMF-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Liquid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/404019

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/FS-2132_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart