KEY466134163 Key Organics / BIONET

(2R)-2-amino-3-(3-chlorophenyl)propanoic acid

  • MDL number MFCD01631984
  • Cas Number : 80126-52-9
  • Molecular Weight : 199.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2R)-2-amino-3-(3-chlorophenyl)propanoic acid
Inchi Code 1S/C9H10ClNO2/c10-7-3-1-2-6(4-7)5-8(11)9(12)13/h1-4,8H,5,11H2,(H,12,13)/t8-/m1/s1
Inchi Key JJDJLFDGCUYZMN-MRVPVSSYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/404338

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/FS-2458_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart