KEY466134168 Key Organics / BIONET

(3,5-dimethoxyphenyl)methanamine

  • MDL number MFCD00052813
  • Cas Number : 34967-24-3
  • Molecular Weight : 167.208

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/405073

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/FS-3204_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart