KEY466134170 Key Organics / BIONET

2-hydroxy-5-(trifluoromethyl)benzonitrile

  • MDL number MFCD11213679
  • Cas Number : 142167-36-0
  • Molecular Weight : 187.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp Solid
Iupac Name 2-hydroxy-5-(trifluoromethyl)benzonitrile
Inchi Code 1S/C8H4F3NO/c9-8(10,11)6-1-2-7(13)5(3-6)4-12/h1-3,13H
Inchi Key FGLWZZOKMAMVSE-UHFFFAOYSA-N

Description

Melting Point Solid
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/405129

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/FS-3263_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart