KEY466134174 Key Organics / BIONET

1-acetyl-2,3-dihydro-1H-indol-3-one

  • MDL number MFCD00466593
  • Cas Number : 16800-68-3
  • Molecular Weight : 175.187

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/405176

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H317,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/FS-3314_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart