KEY466134542 Key Organics / BIONET

2-(furan-2-yl)-1,3-thiazolidine-4-carboxylic acid

  • MDL number MFCD00170602
  • Cas Number : 72678-98-9
  • Molecular Weight : 199.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(2-furyl)-1,3-thiazolidine-4-carboxylic acid
Inchi Code 1S/C8H9NO3S/c10-8(11)5-4-13-7(9-5)6-2-1-3-12-6/h1-3,5,7,9H,4H2,(H,10,11)
Inchi Key VYGBVPOHBURDGB-UHFFFAOYSA-N

Description

Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/429097

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/MS-6498_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart