KEY466135775 Key Organics / BIONET

2-amino-5-phenyl-4,5,6,7-tetrahydro-1,3-benzothiazol-7-one

  • MDL number MFCD00170019
  • Cas Number : 334506-28-4
  • Molecular Weight : 244.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-imino-5-phenyl-2,3,5,6-tetrahydro-1,3-benzothiazol-7(4H)-one
Inchi Code 1S/C13H12N2OS/c14-13-15-10-6-9(7-11(16)12(10)17-13)8-4-2-1-3-5-8/h1-5,9H,6-7H2,(H2,14,15)
Inchi Key HRSUKZMYFXBXMT-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/430382

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/MS-9152_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart