KEY466135816 Key Organics / BIONET

2-aminopyridin-4-ol

  • MDL number MFCD04114161
  • Cas Number : 33631-05-9
  • Molecular Weight : 110.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-amino-4(1H)-pyridinone
Inchi Code 1S/C5H6N2O/c6-5-3-4(8)1-2-7-5/h1-3H,(H3,6,7,8)
Inchi Key HQNIMNQVKVPZES-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/459364

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/TS-01534_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart