KEY466135839 Key Organics / BIONET

2-(3-fluoro-4-methylphenyl)acetonitrile

  • MDL number MFCD01631535
  • Cas Number : 261951-73-9
  • Molecular Weight : 149.168

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Oil
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/459774

Safety Information

signalword Danger
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H301,H311,H331
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/TS-01955_SDS.pdf
pictograms GHS06
Availability details for
Quantity
  • Availability
  • Add to Cart