KEY466135879 Key Organics / BIONET

1-(4-amino-3-bromophenyl)ethan-1-one

  • MDL number MFCD00272386
  • Cas Number : 56759-32-1
  • Molecular Weight : 214.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4-amino-3-bromophenyl)ethanone
Inchi Code 1S/C8H8BrNO/c1-5(11)6-2-3-8(10)7(9)4-6/h2-4H,10H2,1H3
Inchi Key ASMVJBACZFHISI-UHFFFAOYSA-N

Description

Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/460843

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/TS-03054_SDS.pdf
hcodes H302,H312,H315,H317,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart