KEY466135884 Key Organics / BIONET

6-(4-fluorophenyl)pyridine-3-carboxylic acid

  • MDL number MFCD06410388
  • Cas Number : 223127-24-0
  • Molecular Weight : 217.199

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Solid
Purity 97
Storage temp. Store at -18°C
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/460895

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404,P411
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/TS-03107_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart