KEY466135886 Key Organics / BIONET

methyl 3-amino-4-bromobenzoate

  • MDL number MFCD00553090
  • Cas Number : 46064-79-3
  • Molecular Weight : 230.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 3-amino-4-bromobenzoate
Inchi Code 1S/C8H8BrNO2/c1-12-8(11)5-2-3-6(9)7(10)4-5/h2-4H,10H2,1H3
Inchi Key GPMSTDWXXKAKCT-UHFFFAOYSA-N

Description

Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/461057

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/TS-03273_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart