KEY491462635 Key Organics / BIONET

2-methyl-9H-fluorene

  • MDL number MFCD00040873
  • Cas Number : 1430-97-3
  • Molecular Weight : 180.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-methyl-9H-fluorene
Inchi Code 1S/C14H12/c1-10-6-7-14-12(8-10)9-11-4-2-3-5-13(11)14/h2-8H,9H2,1H3
Inchi Key RKJHJMAZNPASHY-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/651387

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-3223_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart