KEY491462650 Key Organics / BIONET

1-{2-[4-(chloromethyl)phenoxy]ethyl}azepane hydrochloride

  • MDL number MFCD11865353
  • Cas Number : 223251-25-0
  • Molecular Weight : 304.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-{2-[4-(chloromethyl)phenoxy]ethyl}azepane hydrochloride
Inchi Code 1S/C15H22ClNO.ClH/c16-13-14-5-7-15(8-6-14)18-12-11-17-9-3-1-2-4-10-17;/h5-8H,1-4,9-13H2;1H
Inchi Key RMVQWASPAGLTRZ-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/651847

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4934_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart