KEY497445046 Key Organics / BIONET

tert-butyl N-(4-bromo-2,6-dinitrophenyl)-N-[(tert-butoxy)carbonyl]carbamate

  • MDL number MFCD29090743
  • Cas Number : 1929606-71-2
  • Molecular Weight : 462.253

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Solid
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/654952

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-3230_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart